Anti-CXCL11 Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8297-99EA 3240 DKK
USBIC8297-99
Anti-CXCL11 Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: CXCL11 (chemokine (C-X-C motif) ligand 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR