Anti-CXCL9 Mouse Monoclonal Antibody [clone: 1F5]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8297-98WEA 3500 DKK
USBIC8297-98W
Anti-CXCL9 Mouse Monoclonal Antibody [clone: 1F5]
Antistoffer
Type: Primary
Antigen: CXCL9 (chemokine (C-X-C motif) ligand 9)
Clonality: Monoclonal
Clone: 1F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR