Anti-CXCL5 Mouse Monoclonal Antibody [clone: 2A9]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8297-98H2EA 5630 DKK
USBIC8297-98H2
Anti-CXCL5 Mouse Monoclonal Antibody [clone: 2A9]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CXCL5 (chemokine (C-X-C motif) ligand 5)
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR