Anti-CXCL5 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8297-98HEA 4750 DKK
USBIC8297-98H
Anti-CXCL5 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 0.5-2?g/ml; Western Blot: 0.1-0.2?g/ml; Functional Assays: 15-20 ?g/ml

Type: Primary
Antigen: CXCL5 (chemokine (C-X-C motif) ligand 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR