Anti-CUX1 Mouse Monoclonal Antibody [clone: 10B2624]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8220EA 3500 DKK
USBIC8220
Anti-CUX1 Mouse Monoclonal Antibody [clone: 10B2624]
Antistoffer
Recommended dilution: Immunochemistry: 1. 5 ?g/ml; Western Blot: 1:500-1:1000; ELISA: 1 ?g/ml-3ng/ml

Type: Primary
Antigen: CUX1 (cut-like homeobox 1)
Clonality: Monoclonal
Clone: 10B2624
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR