Anti-CSE1L Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2614]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8049-88C-APEA 0 DKK
USBIC8049-88C-AP
Anti-CSE1L Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2614]
Type: Primary
Antigen: CSE1L (CSE1 chromosome segregation 1-like (yeast))
Clonality: Monoclonal
Clone: 10B2614
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR