Anti-C-type Natriuretic Peptide-22 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7979-90EA 5820 DKK
USBIC7979-90
Anti-C-type Natriuretic Peptide-22 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1600

Type: Primary
Antigen: C-type Natriuretic Peptide-22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR