Anti-CTGFL Mouse monoclonal antibody [clone: 9E3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7978-35BEA 2300 DKK
USBIC7978-35B
Anti-CTGFL Mouse monoclonal antibody [clone: 9E3]
Antistoffer
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.1-1 ?g/ml

Type: Primary
Antigen: CTGFL
Clonality: Monoclonal
Clone: 9000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR