Anti-CTGF Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7978-25FEA 4750 DKK
USBIC7978-25F
Anti-CTGF Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 0.5-2?g/ml; Western Blot: 0.1-0.2?g/ml; Functional Assays: 6-9?g/ml

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR