Anti-CTGF Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7978-25D-50EA 2900 DKK
USBIC7978-25D-50
Anti-CTGF Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 0.25-1.0 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR