Anti-CTGF Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7978-25BEA 3700 DKK
USBIC7978-25B
Anti-CTGF Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR