Anti-CUL7 Rabbit Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7949-22FEA 5870 DKK
USBIC7949-22F
Anti-CUL7 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:100; Western Blot: 1:1000
Type: Primary
Antigen: CUL7 (cullin 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human