Anti-CUL4B Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7949-21L1EA 3290 DKK
USBIC7949-21L1
Anti-CUL4B Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot:1 ?g/ml; Immunochemistry:2?g/ml

Type: Primary
Antigen: CUL4B (cullin 4B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Xenopus laevis, Zebrafish
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR