Anti-CUL3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7949-21CEA 4730 DKK
USBIC7949-21C
Anti-CUL3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000; Immunochemistry: 1:500

Type: Primary
Antigen: CUL3 (cullin 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR