Anti-CUL2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7949-10DEA 5360 DKK
USBIC7949-10D
Anti-CUL2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:100; Western Blot: 1:1000

Type: Primary
Antigen: CUL2 (cullin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR