Anti-CUL1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7949-06AEA 4730 DKK
USBIC7949-06A
Anti-CUL1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000; Immunochemistry: 1:500

Type: Primary
Antigen: CUL1 (cullin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR