Anti-CUL1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7949-01GEA 5360 DKK
USBIC7949-01G
Anti-CUL1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:50

Type: Primary
Antigen: CUL1 (cullin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR