Anti-CELF5 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7948-53EA 3460 DKK
USBIC7948-53
Anti-CELF5 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: CELF5 (CUGBP, Elav-like family member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR