Anti-C1QTNF6 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7948-25VEA 3590 DKK
USBIC7948-25V
Anti-C1QTNF6 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 10 ?g/ml; Western Blot: 1-2?g/ml

Type: Primary
Antigen: C1QTNF6 (C1q and tumor necrosis factor related protein 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR