Anti-C1QTNF4 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7948-24CEA 3930 DKK
USBIC7948-24C
Anti-C1QTNF4 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: C1QTNF4 (C1q and tumor necrosis factor related protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR