Anti-C1QTNF2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7948-23AEA 5140 DKK
USBIC7948-23A
Anti-C1QTNF2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 2-4?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: C1QTNF2 (C1q and tumor necrosis factor related protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR