Anti-C1QTNF1 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2623]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7948-18D-HRPEA 0 DKK
USBIC7948-18D-HRP
Anti-C1QTNF1 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2623]
Type: Primary
Antigen: C1QTNF1 (C1q and tumor necrosis factor related protein 1)
Clonality: Monoclonal
Clone: 10B2623
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR