Anti-C1QTNF1 Mouse Monoclonal Antibody [clone: 7H203]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7948-18EA 4320 DKK
USBIC7948-18
Anti-C1QTNF1 Mouse Monoclonal Antibody [clone: 7H203]
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: C1QTNF1 (C1q and tumor necrosis factor related protein 1)
Clonality: Monoclonal
Clone: 7H203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR