Anti-SLC31A1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7948-17EA 3500 DKK
USBIC7948-17
Anti-SLC31A1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:250

Type: Primary
Antigen: SLC31A1 (solute carrier family 31 (copper transporter), member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Pig, Rat, Xenopus laevis, Zebrafish
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR