Anti-CTDSP2 Mouse monoclonal antibody [clone: 8C88]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7947-95EA 2770 DKK
USBIC7947-95
Anti-CTDSP2 Mouse monoclonal antibody [clone: 8C88]
Antistoffer
Type: Primary
Antigen: CTDSP2 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2)
Clonality: Monoclonal
Clone: 8C88
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR