Anti-CTBS Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2310]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7947-28B-APEA 0 DKK
USBIC7947-28B-AP
Anti-CTBS Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2310]
Type: Primary
Antigen: CTBS (chitobiase, di-N-acetyl-)
Clonality: Monoclonal
Clone: 10B2310
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR