Anti-CTBS Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7947-28EA 4780 DKK
USBIC7947-28
Anti-CTBS Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: CTBS (chitobiase, di-N-acetyl-)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR