Anti-CTBP-1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2621]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7947-01D-FITCEA 0 DKK
USBIC7947-01D-FITC
Anti-CTBP-1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2621]
Type: Primary
Antigen: CTBP-1
Clonality: Monoclonal
Clone: 10B2621
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR