Anti-CTBP-1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7947-01CEA 3890 DKK
USBIC7947-01C
Anti-CTBP-1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunofluorescence: 1 ?g/ml

Type: Primary
Antigen: CTBP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR