Anti-CSTF3 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2620]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7946-70L-APCEA 0 DKK
USBIC7946-70L-APC
Anti-CSTF3 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2620]
Type: Primary
Antigen: CSTF3 (cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa)
Clonality: Monoclonal
Clone: 10B2620
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR