Anti-CSN2 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2308]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7945-60M-PEEA 0 DKK
USBIC7945-60M-PE
Anti-CSN2 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2308]
Type: Primary
Antigen: CSN2 (casein beta)
Clonality: Monoclonal
Clone: 10B2308
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR