Anti-COPS5 Mouse Monoclonal Antibody [clone: 8H8.5]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7945-54CEA 4320 DKK
USBIC7945-54C
Anti-COPS5 Mouse Monoclonal Antibody [clone: 8H8.5]
Antistoffer
Recommended dilution: Immunochemistry: 1:100

Type: Primary
Antigen: COPS5 (COP9 signalosome subunit 5)
Clonality: Monoclonal
Clone: 8H8.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR