Anti-COPS3 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7945-52CEA 4120 DKK
USBIC7945-52C
Anti-COPS3 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:4000; Western Blot: 1-2?g/ml

Type: Primary
Antigen: COPS3 (COP9 signalosome subunit 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR