Anti-Csk Binding Protein Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7945-16HEA 3410 DKK
USBIC7945-16H
Anti-Csk Binding Protein Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: Csk Binding Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR