Anti-CSK Goat polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7945-05DEA 4120 DKK
USBIC7945-05D
Anti-CSK Goat polyclonal antibody
Antistoffer
Type: Primary
Antigen: CSK (c-src tyrosine kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat