Anti-CSK Mouse monoclonal antibody [clone: 6A399]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7945-05BEA 4510 DKK
USBIC7945-05B
Anti-CSK Mouse monoclonal antibody [clone: 6A399]
Antistoffer
Recommended dilution: Western Blot: 1:125-1:150

Type: Primary
Antigen: CSK (c-src tyrosine kinase)
Clonality: Monoclonal
Clone: 6A399
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR