Anti-CSK Mouse monoclonal antibody [clone: CSK-04]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7945-02EEA 5540 DKK
USBIC7945-02E
Anti-CSK Mouse monoclonal antibody [clone: CSK-04]
Antistoffer
Recommended dilution: Western Blot: 1:250-1:50

Type: Primary
Antigen: CSK (c-src tyrosine kinase)
Clonality: Monoclonal
Clone: CSK-04
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR