Anti-CSGALNACT2 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-97-BIOTEA 6360 DKK
USBIC7944-97-BIOT
Anti-CSGALNACT2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: CSGALNACT2 (chondroitin sulfate N-acetylgalactosaminyltransferase 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR