Anti-CSH1 Chicken polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-96EA 3240 DKK
USBIC7944-96
Anti-CSH1 Chicken polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:500; Immunochemistry: 1:200

Type: Primary
Antigen: CSH1 (chorionic somatomammotropin hormone 1 (placental lactogen))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR