Anti-CRYGD Mouse monoclonal antibody [clone: 10B2608]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-88EA 0 DKK
USBIC7944-88
Anti-CRYGD Mouse monoclonal antibody [clone: 10B2608]
Type: Primary
Antigen: CRYGD (crystallin, gamma D)
Clonality: Monoclonal
Clone: 10B2608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR