Anti-Crystallin, beta Mouse monoclonal antibody [clone: 4E56]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-77-200EA 5340 DKK
USBIC7944-77-50 USBIC7944-77-200
Anti-Crystallin, beta Mouse monoclonal antibody [clone: 4E56]
Antistoffer
Type: Primary
Antigen: Crystallin, beta
Clonality: Monoclonal
Clone: 4E+56
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR