Anti-CRYAB Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-75F-200EA 7610 DKK
USBIC7944-75F-200 USBIC7944-75F-50
Anti-CRYAB Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Dog, Guinea pig, Hamster, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR