Anti-Crystallin, alpha Rabbit polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7944-74G-200EA 4260 DKK
USBIC7944-74G-200
Anti-Crystallin, alpha Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:5000-1:10000
Type: Primary
Antigen: Crystallin, alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Mouse, Rat