Anti-CRYAA Mouse monoclonal antibody [clone: 10B2610]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-72CEA 4260 DKK
USBIC7944-72C
Anti-CRYAA Mouse monoclonal antibody [clone: 10B2610]
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: CRYAA (crystallin, alpha A)
Clonality: Monoclonal
Clone: 10B2610
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR