Anti-CRYAB Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-60BEA 3180 DKK
USBIC7944-60B
Anti-CRYAB Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR