Anti-CRYAA Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-50-200EA 4730 DKK
USBIC7944-50-200 USBIC7944-50-50
Anti-CRYAA Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: CRYAA (crystallin, alpha A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR