Anti-Cryptosporidium sp. Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B2304]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-22BEA 4190 DKK
USBIC7944-22B
Anti-Cryptosporidium sp. Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B2304]
Antistoffer
Recommended dilution: Flow Cytometry: 1:5-1:50;

Type: Primary
Antigen: Cryptosporidium sp.
Clonality: Monoclonal
Clone: 10B2304
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR