Anti-Cryptosporidium parvum Goat Polyclonal Antibody (FITC (Fluorescein))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-16AEA 2900 DKK
USBIC7944-16A
Anti-Cryptosporidium parvum Goat Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Recommended dilution: Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: Cryptosporidium parvum
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR