Anti-Cryptosporidium parvum Mouse monoclonal antibody [clone: 10B2307]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7944-13DEA 4190 DKK
USBIC7944-13D
Anti-Cryptosporidium parvum Mouse monoclonal antibody [clone: 10B2307]
Antistoffer
Recommended dilution: ELISA: 1:20-1:200; Immunofluorescence: 1:10-1:50; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Cryptosporidium parvum
Clonality: Monoclonal
Clone: 10B2307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR