Anti-CRY2 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7942-51AEA 4320 DKK
USBIC7942-51A
Anti-CRY2 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CRY2 (cryptochrome circadian clock 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR